Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proprotein Convertase 7/PCSK7 Antibody (OTI1B8), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP273621R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273621R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273621R Supplier Novus Biologicals Supplier No. NBP273621R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proprotein Convertase 7/PCSK7 Monoclonal specifically detects Proprotein Convertase 7/PCSK7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Proprotein Convertase 7/PCSK7
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B8
Conjugate DyLight 550
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, EC 3.4.21.93, hPC8, LPCFLJ23503, Lymphoma proprotein convertase, PC7FLJ45329, PC8prohormone convertase PC7, Prohormone convertase 7, Proprotein convertase 7, Proprotein convertase 8, proprotein convertase subtilisin/kexin type 7, SPC7proprotein convertase PC7, Subtilisin/kexin-like protease PC7
Gene Symbols PCSK7
Host Species Mouse
Immunogen Full length recombinant protein of human Proprotein Convertase 7/PCSK7 (NP_004707) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9159.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.