Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proprotein Convertase 9/PCSK9 Antibody (106), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB074283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074283 Supplier Novus Biologicals Supplier No. NBP290497AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Mouse samples. It is validated for ELISA.

Specifications

Antigen Proprotein Convertase 9/PCSK9
Applications ELISA
Classification Monoclonal
Clone 106
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Proprotein Convertase 9/PCSK9 (NP_705793.1; Met 1-Gln 694).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 255738
Test Specificity No cross-reactivity in ELISA with: Human PCSK9 Rhesus PCSK9 Human PCSK1
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.