Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propyl chloroformate, 98%

Catalog No. ac245291000
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-61-5 | C4H7ClO2 | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-61-5
Molecular Weight (g/mol) 122.55
InChI Key QQKDTTWZXHEGAQ-UHFFFAOYSA-N
Synonym propyl chloroformate, propyl chlorocarbonate, n-propyl chloroformate, carbonochloridic acid, propyl ester, chloroformicacidpropylester, formic acid, chloro-, propyl ester, unii-a5y2t003ju, n-propylchloroformate, propylchloroformate, chloroformic acid propyl ester
PubChem CID 7998
IUPAC Name propyl carbonochloridate

Specifications

Melting Point -70.0°C
Density 1.0900g/mL
Boiling Point 106.0°C
Flash Point 16°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7ClO2
Refractive Index 1.4030 to 1.4050
Linear Formula CH3(CH2)2OCOCl
Quantity 100 g
Specific Gravity 1.09
Merck Index 15,7961
Solubility Information Solubility in water: reacts
SMILES CCCOC(=O)Cl
Viscosity 0.70 mPa.s (20°C)
Formula Weight 122.55
Percent Purity 98%
Chemical Name or Material Propyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 203-687-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.