Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PROSER3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17044720UL
Description
PROSER3 Polyclonal specifically detects PROSER3 in Human samples. It is validated for Western Blot.Specifications
C19orf55 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
chromosome 19 open reading frame 55 | |
Rabbit | |
45 kDa | |
20 μL | |
Primary | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
C19ORF55 | |
Synthetic peptides corresponding to C19orf55 (chromosome 19 open reading frame 55) The peptide sequence was selected from the middle region of C19orf55)(50ug). Peptide sequence IKQNLHTWNSSLLDLETLSLQSRAARLLKRSKASISSSSSLSPSDASTSS. | |
Affinity Purified | |
RUO | |
148137 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction