Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROSER3 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen C19orf55
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17044720
SDP
View Documents
Novus Biologicals
NBP17044720UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP170447
SDP
View Documents
Novus Biologicals
NBP170447
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

PROSER3 Polyclonal specifically detects PROSER3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

C19orf55
Polyclonal
Rabbit
chromosome 19 open reading frame 55
C19ORF55
IgG
45 kDa
Western Blot
Unconjugated
RUO
148137
Synthetic peptides corresponding to C19orf55 (chromosome 19 open reading frame 55) The peptide sequence was selected from the middle region of C19orf55)(50ug). Peptide sequence IKQNLHTWNSSLLDLETLSLQSRAARLLKRSKASISSSSSLSPSDASTSS.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.