Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prostaglandin E Synthase 2/PTGES2 Antibody (OTI2C3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N273623A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273623A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273623A700 Supplier Novus Biologicals Supplier No. NBP273623AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Prostaglandin E Synthase 2/PTGES2 Monoclonal specifically detects Prostaglandin E Synthase 2/PTGES2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Prostaglandin E Synthase 2/PTGES2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2C3
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C9orf15, chromosome 9 open reading frame 15, EC 5.3.99.3, FLJ14038, gamma-interferon-activated transcriptional element-binding factor 1, GATE-binding factor 1, GBF1, membrane-associated prostaglandin E synthase 2, Microsomal prostaglandin E synthase 2, microsomal prostaglandin E synthase-2, mPGES-2, PGES2MGC11289, prostaglandin E synthase 2
Gene Symbols PTGES2
Host Species Mouse
Immunogen Full length recombinant protein of human Prostaglandin E Synthase 2/PTGES2 (NP_079348) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 80142.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.