Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody

Employ part of a comprehensive antibody portfolio Epredia™ Prostate Specific Antigen (PSA), Rabbit Monoclonal Antibody. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

$474.00 - $2316.00

Specifications

Antigen Prostate Specific Antigen (PSA)
Clone EP1588Y
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RM2104S
View Documents
Epredia™
RM2104S
Concentrated 1 mL
Each for $2,316.00
Only null left
Add to Cart
 
RM2104R7
View Documents
Epredia™
RM2104R7
Prediluted 7 mL
Each for $474.00
Only null left
Add to Cart
 
RM2104S0
View Documents
Epredia™
RM2104S0
Concentrated 100 μL N/A N/A N/A
RM2104R1
View Documents
Epredia™
RM2104R1
Prediluted 1 mL N/A N/A N/A
RM2104RQ
View Documents
Epredia™
RM2104RQ
Ready-to-Use 12 mL N/A N/A N/A
Description

Description

PSA is a chymotrypsin-like serine protease (kallikrein family) produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. PSA is used to confirm prostatic acinar cell origin in primary and metastatic carcinoma and to rule out non-prostatic carcinoma mimics.

Host Species: Rabbit

Clone: EP1588Y

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues near the C-terminus of human PSA

Molecular Weight: 34kDa

Positive Control: Normal prostate or prostate carcinoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Prostate Specific Antigen (PSA)
Immunohistochemistry (Paraffin)
Unconjugated
ASR
A synthetic peptide corresponding to residues near the C-terminus of human PSA
EP1588Y
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.