Learn More
Prostatic Acid Phosphatase/ACPP Rabbit anti-Human, Clone: 4, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Prostatic Acid Phosphatase/ACPP |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 4 |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | acid phosphatase, prostate, ACP3, ACP-3, EC 3.1.3.2, PAP, prostatic acid phosphatase, prostatic acid phosphotase |
| Host Species | Rabbit |
| Immunogen | Recombinant Human Prostatic Acid Phosphatase/ACPP protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.