Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 19S S7 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB042458 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042458 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042458 Supplier Novus Biologicals Supplier No. NBP297271AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteasome 19S S7 Polyclonal specifically detects Proteasome 19S S7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Proteasome 19S S7
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 26S protease regulatory subunit 7, 26S proteasome AAA-ATPase subunit RPT1, MGC3004, MSS1ATPase, 2, proteasome (prosome, macropain) 26S subunit, ATPase, 2, Protein MSS1, putative protein product of Nbla10058
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Proteasome 19S S7 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5701
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.