Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 20S alpha 3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB315557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB315557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB315557 Supplier Novus Biologicals Supplier No. NBP297026APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteasome 20S alpha 3 Polyclonal antibody specifically detects Proteasome 20S alpha 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Proteasome 20S alpha 3
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation PBS
Gene Alias EC 3.4.25.1, HC8MGC32631, Macropain subunit C8, MGC12306, Multicatalytic endopeptidase complex subunit C8, proteasome (prosome, macropain) subunit, alpha type, 3, Proteasome component C8, proteasome subunit alpha type-3, proteasome subunit C8, PSC3, PSC8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Proteasome 20S alpha 3 (Accession#: AAH29402.1; Met1-Met255)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5684
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.