Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 20S alpha 3 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB112293 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB112293 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB112293 Supplier Novus Biologicals Supplier No. NBP297026IR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteasome 20S alpha 3 Polyclonal specifically detects Proteasome 20S alpha 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Proteasome 20S alpha 3
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.25.1, HC8MGC32631, Macropain subunit C8, MGC12306, Multicatalytic endopeptidase complex subunit C8, proteasome (prosome, macropain) subunit, alpha type, 3, Proteasome component C8, proteasome subunit alpha type-3, proteasome subunit C8, PSC3, PSC8
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Proteasome 20S alpha 3 (AAH29402.1; Met1-Met255).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5684
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.