Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Proteasome 20S alpha 3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Proteasome 20S alpha 3 Polyclonal specifically detects Proteasome 20S alpha 3 in Human samples. It is validated for Western Blot.Specifications
Proteasome 20S alpha 3 | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
EC 3.4.25.1, HC8MGC32631, Macropain subunit C8, MGC12306, Multicatalytic endopeptidase complex subunit C8, proteasome (prosome, macropain) subunit, alpha type, 3, Proteasome component C8, proteasome subunit alpha type-3, proteasome subunit C8, PSC3, PSC8 | |
PSMA3 | |
IgG | |
This product is specific to Subunit or Isoform: alpha type-3. |
Western Blot | |
Unconjugated | |
RUO | |
P25788-2 | |
5684 | |
Synthetic peptides corresponding to PSMA3(proteasome (prosome, macropain) subunit, alpha type, 3) The peptide sequence was selected from the middle region of PSMA3. Peptide sequence VKDKAFELELSWVGELTNGRHEIVPKDIREEAEKYAKESLKEEDESDDDN. | |
Primary | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title