Learn More
Proteasome 20S alpha 5 Antibody (AH1.1), mFluor Violet 610 SE, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Proteasome 20S alpha 5 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | AH1.1 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.25.1, FLJ42315, macropain subunit zeta, Macropain zeta chain, MGC117302, MGC125802, MGC125803, MGC125804, Multicatalytic endopeptidase complex zeta chain, proteasome (prosome, macropain) subunit, alpha type, 5, proteasome alpha 5 subunit, proteasome component 5, proteasome subunit alpha type-5, proteasome subunit zeta, Proteasome zeta chain, PSC5, ZETA |
| Host Species | Mouse |
| Immunogen | Sub-unit 5a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.