Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 20S alpha 6 Antibody (OTI3B8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273642A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273642A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273642A70 Supplier Novus Biologicals Supplier No. NBP273642AF750

Mouse Monoclonal Antibody

Proteasome 20S alpha 6 Monoclonal specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Proteasome 20S alpha 6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B8
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota
Gene Symbols PSMA6
Host Species Mouse
Immunogen Full length recombinant protein of human Proteasome 20S alpha 6 (NP_002782) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5687.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.