Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 20S alpha 6 Antibody (OTI3B8), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301806 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301806 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301806 Supplier Novus Biologicals Supplier No. NBP273642APC

Mouse Monoclonal Antibody

Proteasome 20S alpha 6 Monoclonal antibody specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Proteasome 20S alpha 6
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3B8
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota
Host Species Mouse
Immunogen Full length human recombinant protein of human PSMA6 (NP_002782) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5687
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.