Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 20S alpha 6 Rabbit anti-Human, Mouse, Rat, Clone: S01-4I2, Novus Biologicals™
SDP

Catalog No. NB188521 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188521 100 μg
NB188522 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188521 Supplier Novus Biologicals Supplier No. NBP315085100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Proteasome 20S alpha 6 Monoclonal antibody specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen Proteasome 20S alpha 6
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone S01-4I2
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota
Host Species Rabbit
Immunogen A synthetic peptide of human Proteasome 20S alpha 6 (Uniprot#: P60900)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5687
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.