Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S alpha 6 Rabbit anti-Human, Mouse, Rat, Clone: S01-4I2, Novus Biologicals™

Rabbit Monoclonal Antibody
$275.50 - $584.50
Specifications
Antigen | Proteasome 20S alpha 6 |
---|---|
Clone | S01-4I2 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
Proteasome 20S alpha 6 Monoclonal antibody specifically detects Proteasome 20S alpha 6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Proteasome 20S alpha 6 | |
Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
5687 | |
IgG | |
Affinity-purified |
S01-4I2 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 3.4.25.1, IOTA, Macropain iota chain, macropain subunit iota, MGC22756, MGC2333, MGC23846, Multicatalytic endopeptidase complex iota chain, P27K, p27KPROS-27, PROS2727 kDa prosomal protein, prosomal P27K protein, proteasome (prosome, macropain) subunit, alpha type, 6, Proteasome iota chain, proteasome subunit alpha type-6, proteasome subunit iota | |
A synthetic peptide of human Proteasome 20S alpha 6 (Uniprot#: P60900) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title