Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S beta2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15459020UL
Description
Proteasome 20S beta2 Polyclonal specifically detects Proteasome 20S beta2 in Human, Mouse samples. It is validated for Western Blot.Specifications
Proteasome 20S beta2 | |
Polyclonal | |
Western Blot 0.2 ug/ml | |
P49721 | |
PSMB2 | |
Synthetic peptides corresponding to PSMB2(proteasome (prosome, macropain) subunit, beta type, 2) The peptide sequence was selected from the middle region of PSMB2 (NP_002785). Peptide sequence LDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLD. | |
Affinity Purified | |
RUO | |
5690 | |
Human, Mouse | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2 | |
Rabbit | |
23 kDa | |
20 μL | |
Primary | |
This product is specific to Subunit or Isoform: beta type-2. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction