Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proteasome 20S beta2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Proteasome 20S beta2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15459020
![]() |
Novus Biologicals
NBP15459020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154590
![]() |
Novus Biologicals
NBP154590 |
100 μL |
Each for $487.50
|
|
|||||
Description
Proteasome 20S beta2 Polyclonal specifically detects Proteasome 20S beta2 in Human samples. It is validated for Western Blot.Specifications
Proteasome 20S beta2 | |
Polyclonal | |
Rabbit | |
P49721 | |
5690 | |
Synthetic peptides corresponding to PSMB2(proteasome (prosome, macropain) subunit, beta type, 2) The peptide sequence was selected from the middle region of PSMB2 (NP_002785). Peptide sequence LDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLD. | |
Primary | |
23 kDa |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.4.25.1, HC7-I, Macropain subunit C7-I, MGC104215, MGC126885, multicatalytic endopeptidase complex subunit C7-1, Multicatalytic endopeptidase complex subunit C7-I, proteasome (prosome, macropain) subunit, beta type, 2, proteasome beta 2 subunit, Proteasome component C7-I, proteasome subunit beta type-2, proteasome subunit, beta type, 2 | |
PSMB2 | |
IgG | |
This product is specific to Subunit or Isoform: beta type-2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title