Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome 26S S5 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB344708 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344708 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344708 Supplier Novus Biologicals Supplier No. NB1203319APC

Rabbit Polyclonal Antibody

Proteasome 26S S5 Polyclonal antibody specifically detects Proteasome 26S S5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Proteasome 26S S5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias KIAA007226S proteasome non-ATPase regulatory subunit 5, proteasome (prosome, macropain) 26S subunit, non-ATPase, 5,26S proteasome subunit S5B, S5BMGC23145,26S protease subunit S5 basic
Host Species Rabbit
Immunogen Synthetic Peptide: L(485) S E G P Y Y V K P V S T T A V E G A E(504)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5711
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.