Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome beta 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP154583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154583 Supplier Novus Biologicals Supplier No. NBP154583
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Proteasome beta 1 Polyclonal specifically detects Proteasome beta 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Proteasome beta 1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P20618
Gene Alias EC 3.4.25.1, FLJ25321, HC5, KIAA1838, Macropain subunit C5, Multicatalytic endopeptidase complex subunit C5, PMSB1, proteasome (prosome, macropain) subunit, beta type, 1, proteasome beta 1 subunit, Proteasome component C5, Proteasome gamma chain, proteasome subunit beta type-1, proteasome subunit HC5, PSC5
Gene Symbols PSMB1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PSMB1(proteasome (prosome, macropain) subunit, beta type, 1) The peptide sequence was selected from the middle region of PSMB1 (NP_002784). Peptide sequence NNKAMTTGAIAAMLSTILYSRRFFPYYVYNIIGGLDEEGKGAVYSFDPVG.
Molecular Weight of Antigen 26 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5689
Test Specificity This product is specific to Subunit or Isoform: beta type-1.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.