Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteasome subunit beta type 4 Antibody (OTI2C9), PerCP, Novus Biologicals™
SDP

Catalog No. NB301799 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301799 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301799 Supplier Novus Biologicals Supplier No. NBP273655PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Proteasome subunit beta type 4 Monoclonal antibody specifically detects Proteasome subunit beta type 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Proteasome subunit beta type 4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C9
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias EC 3.4.25.1, HN3, hsBPROS26, HsN3, Macropain beta chain, Multicatalytic endopeptidase complex beta chain, PROS26PROS-26, proteasome (prosome, macropain) subunit, beta type, 4, Proteasome beta chain, Proteasome chain 3, proteasome subunit beta type-4, proteasome subunit HsN3, proteasome subunit, beta type, 4,26 kDa prosomal protein
Host Species Mouse
Immunogen Full length human recombinant protein of human PSMB4(NP_002787) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5692
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.