Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein A Antibody, Novus Biologicals™
SDP

Catalog No. NBP253424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP253424 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP253424 Supplier Novus Biologicals Supplier No. NBP253424
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Protein A Polyclonal specifically detects Protein A in Bacteria samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen Protein A
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:20000-1:100000, Immunoprecipitation 1:1000-1:5000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Alias Immunoglobulin G binding protein A, SPA, Staphylococcal protein A
Gene Symbols SAOUHSC_00069
Host Species Goat
Immunogen Protein A [Staphylococcus aureus]
Purification Method Multi-step
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Epitope Tags, Fusion Proteins, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3919448
Target Species Bacteria
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.