Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7146642
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB99761SP 25 μg
MAB99761100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB99761SP Supplier R&D Systems Supplier No. MAB99761SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Protein A Monoclonal specifically detects Protein A in Multi-Species samples. It is validated for Western Blot.

Specifications

Antigen Protein A
Applications Western Blot
Classification Monoclonal
Clone 946418
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 20 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Immunoglobulin G binding protein A, PROA, Protein-A, SPA, Staphylococcal protein A
Host Species Rat
Immunogen Recombinant Protein A
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3919448.0
Test Specificity Detects recombinant Protein A in direct ELISA and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species All species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.