Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A and Protein G Mag Sepharose™

Catalog No. 45002173 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 μL
4 x 500 μL
Binding Capacity:
13 to 22mg Human IgG/mL
8 to 17mg Human IgG/mL
Ligand:
Native Protein A
Protein G
Bead Type:
Protein A Mag Sepharose
Protein G Mag Sepharose
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Binding Capacity Ligand Bead Type
45-002-173 4 x 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
45-002-161 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
45-002-160 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
45-002-174 4 x 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 45-002-173 Supplier Cytiva Supplier No. 28951379
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simplify enrichment of target proteins by immunoprecipitation or pull-down applications

Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

  • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: Native protein A; Native protein G
Binding capacity Protein A: 8-17mg human IgG/mL gel
Binding capacity Protein G: 13-22mg human IgG/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% ethanol
Storage temperature: 4°C to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Protein G Mag Sepharose
Binding Capacity 13 to 22mg Human IgG/mL
Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Quantity 4 x 500 μL
Content And Storage 4°C to 8°C
For Use With (Application) Designed for covalent coupling of any Protein or biomolecule containing a free Amine group and is used for various types of pull down experiments, for biomarker research
Ligand Protein G
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.