Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Disulfide Isomerase/P4HB Antibody (537331), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB303630 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303630 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303630 Supplier Novus Biologicals Supplier No. FAB4236MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein Disulfide Isomerase/P4HB Monoclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen Protein Disulfide Isomerase/P4HB
Applications Immunocytochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 537331
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase, thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, thyroid hormone-binding protein p55
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protein Disulfide Isomerase/P4HB, Asp18-Lys505, Accession # P07237
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cellular Markers, ER Markers, Fibroblast Cell Markers, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 5034
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.