Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Protein G Mag Sepharose™ Xtra
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL
Description
- Non adherant beads
- Matrix: paramagnetic, spherical, highly cross-linked agarose particles
- Ligand: Protein G
- Color: black
- Operating temperature: room temperature
- Can be used without detergents
- For small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants
- Available in 2 x 1mL and 5 x 1mL packs

Specifications
Specifications
Bead Type | Protein G Mag Sepharose Xtra magnetic bead |
Binding Capacity | 27mg human IgG/mL gel |
Matrix | Paramagnetic, spherical, highly cross-linked agarose particles |
Quantity | 2 x 1 mL |
Content And Storage | 4°C to 8°C |
For Use With (Application) | For rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants |
Ligand | Protein G |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction