Learn More
Protein Gene Product 9.5 (PGP9.5) Rabbit Polyclonal Antibody, Epredia™
Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Protein Gene Product 9.5 (PGP9.5), Rabbit Polyclonal Antibody.
Supplier: Epredia RB9202R7
Description
Protein gene product (PGP9.5) is a neuron specific protein, structurally and immunologically distinct from neuron specific enolase. The protein, which has a molecular weight of 27kDa, was first identified by high resolution two dimensional PAGE. Standard immunohistochemical techniques have demonstrated the presence of PGP9.5 in neurons and nerve fibres at all levels of the central and peripheral nervous system, in many neuroendocrine cells, in segments of the renal tubules, in spermatogonia and Leydig cells of the testis, in ova and in some cells of both the pregnant and non-pregnant corpus luteum. This antibody is of value as a marker for neurons and may be of particular use in the study of ubiquitinated cellular inclusions characteristic of several chronic human neurodegenerative diseases. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Recombinant full-length human PGP9.5 protein
Molecular Weight: 9.5kDa
Positive Control: Pancreas and small intestine
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Protein Gene Product 9.5 (PGP9.5) | |
Polyclonal | |
Rabbit | |
7 mL | |
Neuroscience | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Recombinant full-length human PGP95 protein | |
IVD | |
Primary | |
Prediluted |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.