Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Gene Product 9.5 (PGP9.5) Rabbit Polyclonal Antibody, Epredia™

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Protein Gene Product 9.5 (PGP9.5), Rabbit Polyclonal Antibody.

Specifications

Antigen Protein Gene Product 9.5 (PGP9.5)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
RB9202R1
View Documents
Epredia
RB9202R1
Prediluted 1 mL N/A N/A N/A
RB9202R7
View Documents
Epredia
RB9202R7
Prediluted 7 mL N/A N/A N/A
Description

Description

Protein gene product (PGP9.5) is a neuron specific protein, structurally and immunologically distinct from neuron specific enolase. The protein, which has a molecular weight of 27kDa, was first identified by high resolution two dimensional PAGE. Standard immunohistochemical techniques have demonstrated the presence of PGP9.5 in neurons and nerve fibres at all levels of the central and peripheral nervous system, in many neuroendocrine cells, in segments of the renal tubules, in spermatogonia and Leydig cells of the testis, in ova and in some cells of both the pregnant and non-pregnant corpus luteum. This antibody is of value as a marker for neurons and may be of particular use in the study of ubiquitinated cellular inclusions characteristic of several chronic human neurodegenerative diseases. This antibody is suitable for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Recombinant full-length human PGP9.5 protein

Molecular Weight: 9.5kDa

Positive Control: Pancreas and small intestine

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Protein Gene Product 9.5 (PGP9.5)
Polyclonal
Rabbit
Neuroscience
Recombinant full-length human PGP95 protein
Primary
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.