Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Kinase D2 Antibody (OTI5G1), Novus Biologicals™
SDP

Catalog No. NBP202984 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202984 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202984 Supplier Novus Biologicals Supplier No. NBP202984
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein Kinase D2 Monoclonal specifically detects Protein Kinase D2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Protein Kinase D2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5G1
Concentration 0.72 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150
Gene Alias DKFZP586E0820, EC 2.7.11, EC 2.7.11.13, nPKC-D2, PKD2HSPC187, protein kinase D2, serine/threonine-protein kinase D2
Gene Symbols PRKD2
Host Species Mouse
Immunogen Full length human recombinant protein of human PRKD2 (NP_057541) produced in HEK293T cell.
Molecular Weight of Antigen 96.5 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25865
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.