Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Kinase D2 Antibody (OTI5G1), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N27364PECY7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N27364PECY7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N27364PECY7 Supplier Novus Biologicals Supplier No. NBP273664PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein Kinase D2 Monoclonal specifically detects Protein Kinase D2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen Protein Kinase D2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI5G1
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZP586E0820, EC 2.7.11, EC 2.7.11.13, nPKC-D2, PKD2HSPC187, protein kinase D2, serine/threonine-protein kinase D2
Gene Symbols PRKD2
Host Species Mouse
Immunogen Full length recombinant protein of human Protein Kinase D2 (NP_057541) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 25865.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.