Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein O-Fucosyltransferase 1/POFUT1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB356996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356996 Supplier Novus Biologicals Supplier No. NBP176940MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Protein O-Fucosyltransferase 1/POFUT1 Polyclonal antibody specifically detects Protein O-Fucosyltransferase 1/POFUT1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Protein O-Fucosyltransferase 1/POFUT1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FUT12EC 2.4.1.221, GDP-fucose protein O-fucosyltransferase 1, KIAA0180MGC2482, o-fucosyltransferase protein, O-Fuc-T, O-FucT-1, O-FUT, Peptide-O-fucosyltransferase 1, protein O-fucosyltransferase 1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human POFUT1. The immunogen is located within amino acids 90 - 140 of POFUT1. Amino Acid Squence: DFMEKLAPTHWPPEKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23509
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.