Learn More
Protein phosphatase 1F Antibody (6D11), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Protein phosphatase 1F |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 6D11 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F |
| Gene Symbols | PPM1F |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.