Learn More
Description
Specifications
Specifications
| Antigen | Protein phosphatase 1F |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 6D11 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA |
| Formulation | PBS with No Preservative |
| Gene Alias | Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F |
| Gene Symbols | PPM1F |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
