Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein phosphatase 1F Antibody (6D11), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309952 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309952 Supplier Novus Biologicals Supplier No. NBP242648MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein phosphatase 1F Monoclonal antibody specifically detects Protein phosphatase 1F in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Protein phosphatase 1F
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 6D11
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F
Host Species Mouse
Immunogen Recombinant human Protein phosphatase 1F (1-454aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 9647
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.