Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Z Antibody (2B4), Novus Biologicals™
SDP

Catalog No. NBP147452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147452 Supplier Novus Biologicals Supplier No. NBP147452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Protein Z
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B4
Conjugate Unconjugated
Dilution Western Blot 1: 500-1:2000, ELISA 1:10000
Gene Alias protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z
Gene Symbols PROZ
Host Species Mouse
Immunogen Purified recombinant fragment of Protein Z expressed in E. Coli.
Molecular Weight of Antigen 45 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8858
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.