Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Z Antibody (OTI4D5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP273673IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273673IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273673IR Supplier Novus Biologicals Supplier No. NBP273673IR

Mouse Monoclonal Antibody

Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Protein Z
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI4D5
Conjugate DyLight 755
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z
Gene Symbols PROZ
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 115-400 of human Protein Z (NP_003882) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8858.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.