Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proteinase K Lyophilized Powder

Catalog No. aaj76230mc
Click to view available options
Quantity:
100 mg

28.9 KDa

  • Used in the isolation of native, high molecular weight DNA and RNA.
  • Used as a general protease to inactivate DNases, RNases and to degrade proteins.
  • Specifically modifies cell surface proteins and glycoproteins for analysis of membrane structures for protein localization.
  • Produces characteristic protein fragments used in enzyme/protein structure and function studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Lyophilized powder
  • Source: Tritirachium album
  • Concentration: 20 - 50units/mg
  • Molecular weight: 28.9kDa
  • Optimum pH: 4.0 - 12.5
  • Optimum temperature: 65°C
  • Activators: Stimulated by 0.2 - 1% SDS or 1 - 4M urea
  • Inactivated by PMSF(1); Not inactivated by EGTA, thiol reagents or trypsin/chymotrypsin inhibitors

Applications:

  • Used in isolation of native, high molecular weight DNA and RNA
  • Used as general protease to inactivate DNases, RNases and to degrade proteins
  • Specifically modifies cell surface proteins and glycoproteins for analysis of membrane structures for protein localization
  • Produces characteristic protein fragments used in enzyme/protein structure and function studies
TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 to 50 U/mg
Host Species T. album
Content And Storage Dry at 4°C
Quantity 100 mg
Product Type Proteinase K Powder
Molecular Weight (g/mol) 28.9 kDa
Form Lyophilized

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.