Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteinase K (PK) Solution for ReliaPrep™ Large Volume HT gDNA Isolation System

Catalog No. pra5051 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
23 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA5051 23 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRA5051 Supplier Promega Supplier No. A5051
Only null left
Add to Cart
Add to Cart

Proteinase K, produced by the fungus Tritirachium album Limber, is a serine protease that exhibits broad cleavage activity. Proteinase K (PK) Solution is supplied at a concentration of 20mg/mL in 10mM Tris-HCl (pH 7.5), 1mM calcium chloride, 50% glycerol.

  • Stable at Room Temperature and Easy to Use
  • Active over pH range 4.3-12.0 in 0.5% SDS or 1% Triton(R) X-100
  • Retains >80% activity at temperatures up to 60°C
  • No resuspension or thawing before use
  • Cat.# MC5005, MC5008 supplied at a concentration of 20mg/ml

Specifications

Content And Storage 15°C to 30°C
Quantity 23 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.