Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin-1 Antibody (648127), PE, Novus Biologicals™
SDP

Catalog No. NB377372 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377372 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377372 Supplier Novus Biologicals Supplier No. FAB5899P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protocadherin-1 Monoclonal antibody specifically detects Protocadherin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Protocadherin-1
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 648127
Conjugate PE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias cadherin-like 1, Cadherin-like protein 1, FLJ53887, MGC45991, pc42, PC42PCDH42, protocadherin 1, protocadherin 1 (cadherin-like 1), protocadherin 42, protocadherin-1, protocadherin-42
Host Species Mouse
Immunogen E. coli-derived recombinant human Protocadherin-1, Thr58-Asn162, Accession # Q08174
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5097
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.