Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin-19 Antibody (921614), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB346000 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346000 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346000 Supplier Novus Biologicals Supplier No. FAB8626MFV450

Mouse Monoclonal Antibody

Protocadherin-19 Monoclonal antibody specifically detects Protocadherin-19 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Protocadherin-19
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 921614
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EFMR;EIEE9;KIAA1313;PCDH19
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Protocadherin-19, Met1-Ser678, Accession # Q8TAB3
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.