Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Protocadherin alpha 4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159253
Description
Protocadherin alpha 4 Polyclonal specifically detects Protocadherin alpha 4 in Human samples. It is validated for Western Blot.Specifications
Protocadherin alpha 4 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PCDHA4 | |
Synthetic peptides corresponding to PCDHA4(protocadherin alpha 4) The peptide sequence was selected from the N terminal of PCDHA4. Peptide sequence VDRPLQVFHVDVEVRDINDNPPVFPATQKNLSIAESRPLDSRFPLEGASD. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Guinea pig: 92%; Canine: 84%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
CNR1, CNRN1, CRNR1, KIAA0345-like 10, MGC138307, MGC142169, ortholog of mouse CNR1, PCDH-alpha-4, PCDH-ALPHA4, protocadherin alpha 4, protocadherin alpha-4 | |
Rabbit | |
Affinity purified | |
RUO | |
56144 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction