Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin gamma A8 Antibody (RB31662), Novus Biologicals™
SDP

Catalog No. NBP230028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230028 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230028 Supplier Novus Biologicals Supplier No. NBP230028

Rabbit Polyclonal Antibody

Protocadherin gamma A8 Polyclonal specifically detects Protocadherin gamma A8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Protocadherin gamma A8
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Clone RB31662
Concentration 0.32 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:50, Immunohistochemistry-Paraffin 1:10 - 1:50
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. Q9Y5G5
Gene Alias KIAA0327PCDH-GAMMA-A8, PCDH-gamma-A8, protocadherin gamma subfamily A, 8, protocadherin gamma-A8
Gene Symbols PCDHGA8
Host Species Rabbit
Immunogen This PCDHGA8 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 781-808 amino acids from the C-terminal region of human PCDHGA8.
Molecular Weight of Antigen 101.48 kDa
Purification Method Affinity Purified
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9708
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.