Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protocadherin gamma C3 Antibody (926518), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB176394 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176394 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176394 Supplier Novus Biologicals Supplier No. FAB83641J
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protocadherin gamma C3 Monoclonal specifically detects Protocadherin gamma C3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Protocadherin gamma C3
Applications Flow Cytometry
Classification Monoclonal
Clone 926518
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias cadherin-like 2, PC43, PC-43, PCDH2, PCDH-GAMMA-C3, protocadherin 2, protocadherin 43, protocadherin gamma subfamily C, 3, protocadherin gamma-C3, protocadherin-2, Protocadherin-43
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin gamma C3, Met1-Tyr693, Accession # Q9UN70
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 5098
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.