Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protogenin Antibody (OTI2B3), PerCP, Novus Biologicals™
SDP

Catalog No. NB301793 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301793 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301793 Supplier Novus Biologicals Supplier No. NBP273674PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Protogenin Monoclonal antibody specifically detects Protogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Protogenin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2B3
Conjugate PerCP
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), shen-dan
Host Species Mouse
Immunogen Full length human recombinant protein of human PRTG (NP_776175) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283659
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.