Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prox1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018373 0.025 mg
NBP118605 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018373 Supplier Novus Biologicals Supplier No. NBP1186050.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Prox1 Polyclonal specifically detects Prox1 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Prox1
Applications Immunohistochemistry (Frozen), Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:750-1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 5mM PBS (pH 7.2) with No Preservative
Gene Accession No. Q92786
Gene Alias Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
Gene Symbols PROX1
Host Species Rabbit
Immunogen Recombinant human Prox-1
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 5629
Reconstitution Reconstitute in sterile water to a concentration of 0.1-1.0mg/mL.
Target Species Mouse, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.