Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$215.00 - $531.00
Specifications
Antigen | Prox1 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Frozen), Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Prox1 Polyclonal antibody specifically detects Prox1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
Prox1 | |
Immunohistochemistry (Frozen), Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Vision | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
PROX1 | |
IgG | |
Protein A purified |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Q92786 | |
5629 | |
Recombinant human Prox-1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title