Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRP19 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB057335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB057335 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB057335 Supplier Novus Biologicals Supplier No. NBP299020H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRP19 Polyclonal specifically detects PRP19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PRP19
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias hPSO4, NMP200SNEV, Nuclear matrix protein 200, nuclear matrix protein NMP200 related to splicing factor PRP19, pre-mRNA-processing factor 19, PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae), PRP19PRP19/PSO4 homolog (S. cerevisiae), PSO4PRP19/PSO4 homolog, Senescence evasion factor, UBOX4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PRP19.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, GPCR, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27339
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.