Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRP19 Antibody, Novus Biologicals™
SDP

Catalog No. NBP131354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP131354 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP131354 Supplier Novus Biologicals Supplier No. NBP131354
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRP19 Polyclonal specifically detects PRP19 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PRP19
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.39mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UMS4
Gene Alias hPSO4, NMP200SNEV, Nuclear matrix protein 200, nuclear matrix protein NMP200 related to splicing factor PRP19, pre-mRNA-processing factor 19, PRP19/PSO4 pre-mRNA processing factor 19 homolog (S. cerevisiae), PRP19PRP19/PSO4 homolog (S. cerevisiae), PSO4PRP19/PSO4 homolog, Senescence evasion factor, UBOX4
Gene Symbols PRPF19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRP19. The exact sequence is proprietary.
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, GPCR, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 27339
Test Specificity Human and Mouse. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.