Learn More
PRPF38A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PRPF38A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8255
![]() |
Bethyl Laboratories, Inc
A304635AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Bovine and Orangutan
PRPF38A is a protein coding gene involved in pre-mRNA splicing as a component of the spliceosome.Specifications
PRPF38A | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Prp38, RP5-965L7.1, pre-mRNA-splicing factor 38A | |
Between 262 and 312 | |
Primary | |
4° C | |
PRPF38A |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q4FK66, Q8NAV1 | |
230596, 84950 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.